Sulfur trioxide trimethylamine complex 95%

Supplier: Thermo Scientific Chemicals
Danger

L00302-14 L00302-22 L00302-36
CAAAAL00302-14EA 64.15 CAD
CAAAAL00302-14 CAAAAL00302-22 CAAAAL00302-36
Sulfur trioxide trimethylamine complex 95%
Sulphur trioxide trimethylamine complex

Purity: 95% CAS No.: 3162-58-1 Molecular Formula: C3H9NO3S Formula Weight: 139.17 Melting Point: ca 235° dec. Beilstein Reference: 3681759 UN No.: 3261 MDL No.: MFCD00012421

Formula: C₃H₉NO₃S
MW: 139.17 g/mol
Melting Pt: ∼ 235 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012421
CAS Number: 3162-58-1
EINECS: 221-614-7
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR