(Ethoxycarbonylmethyl)dimethylsulfonium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2-Ethoxy-2-oxoethyl)(dimethyl)sulfonium bromide, (2-Ethoxy-2-oxoethyl)(dimethyl)sulphonium bromide
L00266-09
L00266-18
CAAAAL00266-09EA
100.65
CAD
CAAAAL00266-09
CAAAAL00266-18
(Ethoxycarbonylmethyl)dimethylsulfonium bromide 98%
(Ethoxycarbonylmethyl)dimethylsulfonium bromide
Purity: 98%
CAS No.: 5187-82-6
Molecular Formula: CH3CH2O2CCH2S(CH3)2Br
Formula Weight: 229.14
Storage Temperature: 2°C to 8°C
Melting Point: ca 80° dec.
Beilstein Reference: 4161472
MDL No.: MFCD00031604
Formula:
CH₃CH₂O₂CCH₂S(CH₃)₂Br MW: 229.14 g/mol Melting Pt: ∼ 80 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00830557 CAS Number: 5187-82-6 EINECS: 225-968-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...