1-(Heptafluorobutyryl)imidazole 98+%

Supplier: Thermo Scientific Chemicals
Warning

L00221-06 L00221-14
CAAAAL00221-06EA 235.42 CAD
CAAAAL00221-06 CAAAAL00221-14
1-(Heptafluorobutyryl)imidazole 98+%
1-(Heptafluorobutyryl)imidazole

Fieser: 3,148


Purity: 98+% CAS No.: 32477-35-3 Molecular Formula: C7H3F7N2O Formula Weight: 264.10 Storage Temperature: -30°C to -10°C Flash Point: 77°(170°F) Refractive Index: 1.3850 Melting Point: 10-13° Boiling Point: 160-162° Beilstein Reference: 4488026 MDL No.: MFCD00014503

Formula: C₇H₃F₇N₂O
Boiling Pt: 160…162 °C
Melting Pt: 10…13 °C
Density: 1,490
Flash Pt: 77 °C(170 °F)
Storage Temperature: Freezer
MDL Number: MFCD00014503
CAS Number: 32477-35-3
EINECS: 251-063-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR