2-Phenyl-5-oxazolone 97%
Supplier: Thermo Scientific Chemicals
L00194-03
L00194-06
L00194-14
CAAAAL00194-03EA
110.28
CAD
CAAAAL00194-03
CAAAAL00194-06
CAAAAL00194-14
2-Phenyl-5-oxazolone 97%
2-Phenyl-5-oxazolone
Purity: 97%
CAS No.: 1199-01-5
Molecular Formula: C9H7NO2
Formula Weight: 161.16
Storage Temperature: 2°C to 8°C
Melting Point: 89-92°
Beilstein Reference: 128465
MDL No.: MFCD00014517
Formula:
C₉H₇NO₂ Melting Pt: 89…92 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00014517 CAS Number: 1199-01-5 EINECS: 214-840-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...