1,3-Diphenylisobenzofuran 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
2,5-Diphenyl-3,4-benzofuran, 1,3-Diphenyl-isobenzofuran, DIB
L00101-03
L00101-06
CAAAAL00101-03EA
89.11
CAD
CAAAAL00101-03
CAAAAL00101-06
1,3-Diphenylisobenzofuran 97%
1,3-Diphenylisobenzofuran
Fieser: 1,342 2,178 4,209
Purity: 97%
CAS No.: 5471-63-6
Molecular Formula: C20H14O
Formula Weight: 270.33
Storage Temperature: 2°C to 8°C
Melting Point: 131-136°
Beilstein Reference: 199922
MDL No.: MFCD00005931
Formula:
C₂₀H₁₄O Melting Pt: 131…136 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005931 CAS Number: 5471-63-6 EINECS: 226-808-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...