4-(Pyrrolidin-3-yl)morpholine 97%

Supplier: Thermo Scientific Chemicals
Danger

H51002-03 H51002-MD
CAAAAH51002-03EA 339.7 CAD
CAAAAH51002-03 CAAAAH51002-MD
4-(Pyrrolidin-3-yl)morpholine 97%
4-(Pyrrolidin-3-yl)morpholine

Purity: 97% CAS No.: 53617-37-1 Molecular Formula: C8H16N2O Formula Weight: 156.23 UN No.: 2735 MDL No.: MFCD06740352

Formula: C₈H₁₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD06740352
CAS Number: 53617-37-1
UN: 2735
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR