1-(2-Fluorophenyl)-2-propylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

H50999-03 H50999-MD
CAAAAH50999-03EA 404.48 CAD
CAAAAH50999-03 CAAAAH50999-MD
1-(2-Fluorophenyl)-2-propylamine 97%
1-(2-Fluorophenyl)-2-propylamine

Purity: 97% CAS No.: 1716-60-5 Molecular Formula: C9H12FN Formula Weight: 153.20 MDL No.: MFCD01708100

Formula: C₉H₁₂FN
Storage Temperature: Ambient
MDL Number: MFCD01708100
CAS Number: 1716-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR