1-Acetyl-4-(methylamino)piperidine

Supplier: Thermo Scientific Chemicals
Danger

H50985-03 H50985-MD
CAAAAH50985-03EA 363.4 CAD
CAAAAH50985-03 CAAAAH50985-MD
1-Acetyl-4-(methylamino)piperidine
1-Acetyl-4-(methylamino)piperidine

CAS No.: 139062-96-7 Molecular Formula: C8H16N2O Formula Weight: 156.23 UN No.: 3259 MDL No.: MFCD07367773

Formula: C₈H₁₆N₂O
MW: 156.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367773
CAS Number: 139062-96-7
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR