Benzylpyridin-3-ylmethylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

H50978-03 H50978-MD
CAAAAH50978-03EA 357.08 CAD
CAAAAH50978-03 CAAAAH50978-MD
Benzylpyridin-3-ylmethylamine 97%
Benzylpyridin-3-ylmethylamine

Purity: 97% CAS No.: 63361-56-8 Molecular Formula: C13H14N2 Formula Weight: 198.27 MDL No.: MFCD08061098

Formula: C₁₃H₁₄N₂
MW: 198.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00716866
CAS Number: 63361-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR