4-(4-Piperidinyloxy)benzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

H50958-03 H50958-MD
CAAAAH50958-03EA 428.18 CAD
CAAAAH50958-03 CAAAAH50958-MD
4-(4-Piperidinyloxy)benzonitrile 98%
4-(4-Piperidinyloxy)benzonitrile

Purity: 98% CAS No.: 224178-67-0 Molecular Formula: C12H14N2O Formula Weight: 202.26 UN No.: 3439 MDL No.: MFCD07368391

Formula: C₁₂H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD07368391
CAS Number: 224178-67-0
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR