4-(4-Chlorophenoxy)piperidine 98%

Supplier: Thermo Scientific Chemicals
Warning

H50944-03 H50944-MD
CAAAAH50944-03EA 412.38 CAD
CAAAAH50944-03 CAAAAH50944-MD
4-(4-Chlorophenoxy)piperidine 98%
4-(4-Chlorophenoxy)piperidine

CAS No.: 97839-99-1 Molecular Formula: C11H14ClNO Formula Weight: 211.69 MDL No.: MFCD04113544

Formula: C₁₁H₁₄ClNO
Storage Temperature: Ambient
MDL Number: MFCD04113544
CAS Number: 97839-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR