3-(4-Chlorophenoxy)piperidine 97%

Supplier: Thermo Scientific Chemicals
Warning

H50935-03 H50935-MD
CAAAAH50935-03EA 300.2 CAD
CAAAAH50935-03 CAAAAH50935-MD
3-(4-Chlorophenoxy)piperidine 97%
3-(4-Chlorophenoxy)piperidine

Purity: 97% CAS No.: 384346-27-4 Molecular Formula: C11H14ClNO Formula Weight: 211.69 MDL No.: MFCD08061075

Formula: C₁₁H₁₄ClNO
Storage Temperature: Ambient
MDL Number: MFCD08061075
CAS Number: 384346-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR