N-Hydroxypropionamidine 97%

Supplier: Thermo Scientific Chemicals
Warning

H50889-03 H50889-MD
CAAAAH50889-03EA 260.7 CAD
CAAAAH50889-03 CAAAAH50889-MD
N-Hydroxypropionamidine 97%
N-Hydroxypropionamidine

Purity: 97% CAS No.: 29335-36-2 Molecular Formula: C3H8N2O Formula Weight: 88.11 MDL No.: MFCD03426272

Formula: C₃H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD03426272
CAS Number: 29335-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR