tert-Butyl methyl(2-oxoethyl)carbamate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl methyll(2-oxoethyl)carbamate, Methyl(2-oxoethyl)carbamic acid tert-butyl ester, N-Boc-(methylamino)acetaldehyde, N-tert-Butoxycarbonyl-(methylamino)acetaldehyde

H50870-03 H50870-MD
CAAAAH50870-03EA 300.2 CAD
CAAAAH50870-03 CAAAAH50870-MD
tert-Butyl methyl(2-oxoethyl)carbamate 97%
tert-Butyl methyl(2-oxoethyl)carbamate

Purity: 98% CAS No.: 123387-72-4 Molecular Formula: C8H15NO3 Formula Weight: 173.21 Storage Temperature: 2°C to 8°C MDL No.: MFCD08064267

Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08064267
CAS Number: 123387-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR