1-(5-Chloro-2-pyridinyl)piperazine 99%

Supplier: Thermo Scientific Chemicals
Warning

H50862-03 H50862-MD
CAAAAH50862-03EA 304.94 CAD
CAAAAH50862-03 CAAAAH50862-MD
1-(5-Chloro-2-pyridinyl)piperazine 99%
1-(5-Chloro-2-pyridinyl)piperazine

Purity: 99% CAS No.: 87394-65-8 Molecular Formula: C9H12ClN3 Formula Weight: 197.67 MDL No.: MFCD04115049

Formula: C₉H₁₂ClN₃
Storage Temperature: Ambient
MDL Number: MFCD04115049
CAS Number: 87394-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR