1-(3-Chloro-2-pyridinyl)piperazine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50859-03
H50859-MD
CAAAAH50859-03EA
376.04
CAD
CAAAAH50859-03
CAAAAH50859-MD
1-(3-Chloro-2-pyridinyl)piperazine 98%
1-(3-Chloro-2-pyridinyl)piperazine
Purity: 98%
CAS No.: 87394-55-6
Molecular Formula: C9H12ClN3
Formula Weight: 197.67
MDL No.: MFCD08061034
Formula:
C₉H₁₂ClN₃ Storage Temperature: Ambient |
MDL Number:
MFCD08061034 CAS Number: 87394-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...