2-(4-Chlorophenoxy)-6-fluorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

H50847-03 H50847-MD
CAAAAH50847-03EA 344.44 CAD
CAAAAH50847-03 CAAAAH50847-MD
2-(4-Chlorophenoxy)-6-fluorobenzaldehyde 98%
2-(4-Chlorophenoxy)-6-fluorobenzaldehyde

Purity: 98% CAS No.: 902836-82-2 Molecular Formula: C13H8ClFO2 Formula Weight: 250.66 MDL No.: MFCD08061024

Formula: C₁₃H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD08061024
CAS Number: 902836-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR