3-(2-Methylphenoxy)pyrrolidine 95%

Supplier: Thermo Scientific Chemicals
Warning

H50784-03 H50784-MD
CAAAAH50784-03EA 224.36 CAD
CAAAAH50784-03 CAAAAH50784-MD
3-(2-Methylphenoxy)pyrrolidine 95%
3-(2-Methylphenoxy)pyrrolidine

Purity: 95% CAS No.: 46196-54-7 Molecular Formula: C11H15NO Formula Weight: 177.25 MDL No.: MFCD08060984

Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08060984
CAS Number: 46196-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR