3-(2-Oxocyclohexyl)propanenitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

H50722-03 H50722-06
CAAAAH50722-03EA 78.21 CAD
CAAAAH50722-03 CAAAAH50722-06
3-(2-Oxocyclohexyl)propanenitrile 99%
3-(2-Oxocyclohexyl)propanenitrile

Purity: 99% CAS No.: 4594-78-9 Molecular Formula: C9H13NO Formula Weight: 151.21 Boiling Point: 138-142°/10mm UN No.: 3276 MDL No.: MFCD00013797

Formula: C₉H₁₃NO
Boiling Pt: 138…142 °C/10 mm
Density: 1.021
Storage Temperature: Ambient
MDL Number: MFCD00013797
CAS Number: 4594-78-9
EINECS: 224-991-6
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR