3,4-Methylenedioxybenzhydrazide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzo[d][1,3]dioxole-5-carbohydrazide

H50713-03 H50713-06
CAAAAH50713-03EA 74.1 CAD
CAAAAH50713-03 CAAAAH50713-06
3,4-Methylenedioxybenzhydrazide 97%
3,4-Methylenedioxybenzhydrazide

Purity: 97% CAS No.: 22026-39-7 Molecular Formula: C8H8N2O3 Formula Weight: 180.16 Melting Point: 176-178° MDL No.: MFCD00060505

Formula: C₈H₈N₂O₃
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00060505
CAS Number: 22026-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR