6-Morpholino-3-pyridinamine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Morpholinopyridin-3-amine, 5-Amino-2-(4-morpholinyl)pyridine
H50699-03
H50699-MD
CAAAAH50699-03EA
413.96
CAD
CAAAAH50699-03
CAAAAH50699-MD
6-Morpholino-3-pyridinamine 97%
6-Morpholino-3-pyridinamine
Purity: 97%
CAS No.: 52023-68-4
Molecular Formula: C9H13N3O
Formula Weight: 179.22
Melting Point: 127-130°
MDL No.: MFCD00119562
Formula:
C₉H₁₃N₃O MW: 179.22 g/mol Melting Pt: 127…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00119562 CAS Number: 52023-68-4 EINECS: 257-613-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...