2'-Hydroxy-4',5'-dimethylacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

H50685-03 H50685-06
CAAAAH50685-03EA 39.18 CAD
CAAAAH50685-03 CAAAAH50685-06
2'-Hydroxy-4',5'-dimethylacetophenone 98%
2'-Hydroxy-4',5'-dimethylacetophenone

Purity: 98% CAS No.: 36436-65-4 Molecular Formula: C10H12O2 Formula Weight: 164.21 Melting Point: 88-90° MDL No.: MFCD00002306

Formula: C₁₀H₁₂O₂
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00002306
CAS Number: 36436-65-4
EINECS: 253-035-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR