Ethyl-3-tert-butyl-1H-pyrazole-5-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50678-03
H50678-06
CAAAAH50678-03EA
147.41
CAD
CAAAAH50678-03
CAAAAH50678-06
Ethyl-3-tert-butyl-1H-pyrazole-5-carboxylate 97%
Ethyl-3-tert-butyl-1H-pyrazole-5-carboxylate
Purity: 97%
CAS No.: 916791-97-4
Molecular Formula: C10H16N2O2
Formula Weight: 196.25
MDL No.: MFCD00173782
Formula:
C₁₀H₁₆N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00173782 CAS Number: 916791-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...