5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole 95%
Supplier: Thermo Scientific Chemicals
H50677-03
H50677-06
CAAAAH50677-03EA
136.04
CAD
CAAAAH50677-03
CAAAAH50677-06
5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole 95%
5-[4-(Trifluoromethyl)phenyl]-1H-tetrazole
Purity: 95%
CAS No.: 2251-79-8
Molecular Formula: C8H5F3N4
Formula Weight: 214.15
MDL No.: MFCD00052525
Formula:
C₈H₅F₃N₄ Storage Temperature: Ambient |
MDL Number:
MFCD06248886 CAS Number: 2251-79-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...