2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime 97%

Supplier: Thermo Scientific Chemicals
Warning

H50673-03 H50673-MD
CAAAAH50673-03EA 281.24 CAD
CAAAAH50673-03 CAAAAH50673-MD
2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime 97%
2-[4-(1,3,4-Oxadiazol-2-yl)phenoxy]acetamidoxime

Purity: 97% CAS No.: 258521-20-9 Molecular Formula: C10H10N4O3 Formula Weight: 234.22 MDL No.: MFCD01764763

Formula: C₁₀H₁₀N₄O₃
Storage Temperature: Ambient
MDL Number: MFCD01764763
CAS Number: 258521-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR