2-(1H-Pyrazol-3-yl)pyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50670-03
H50670-06
CAAAAH50670-03EA
128.45
CAD
CAAAAH50670-03
CAAAAH50670-06
2-(1H-Pyrazol-3-yl)pyridine 98%
2-(1H-Pyrazol-3-yl)pyridine
Purity: 98%
CAS No.: 75415-03-1
Molecular Formula: C8H7N3
Formula Weight: 145.16
Melting Point: 121-122°
MDL No.: MFCD00115151
Formula:
C₈H₇N₃ MW: 145.16 g/mol Melting Pt: 121…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00115151 CAS Number: 75415-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...