Ethyl (2-bromophenyl)acetate 98%

Supplier: Thermo Scientific Chemicals
Warning

H50630-03 H50630-MD
CAAAAH50630-03EA 194.34 CAD
CAAAAH50630-03 CAAAAH50630-MD
Ethyl (2-bromophenyl)acetate 98%
Ethyl (2-bromophenyl)acetate

Purity: 98% CAS No.: 2178-24-7 Molecular Formula: C10H11BrO2 Formula Weight: 243.10 Melting Point: 35-38° MDL No.: MFCD00051921

Formula: C₁₀H₁₁BrO₂
MW: 243.10 g/mol
Melting Pt: 35…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00051921
CAS Number: 2178-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR