N-(4-Bromo-2,6-dimethylphenyl)-N,N-dimethylamine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50584-03
H50584-06
CAAAAH50584-03EA
161.16
CAD
CAAAAH50584-03
CAAAAH50584-06
N-(4-Bromo-2,6-dimethylphenyl)-N,N-dimethylamine 98%
N-(4-Bromo-2,6-dimethylphenyl)-N,N-dimethylamine
Purity: 98%
CAS No.: 50638-54-5
Molecular Formula: C10H14BrN
Formula Weight: 228.13
MDL No.: MFCD00100113
Formula:
C₁₀H₁₄BrN Storage Temperature: Ambient |
MDL Number:
MFCD00100113 CAS Number: 50638-54-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...