1-Bromo-2-(trifluoromethylthio)benzene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromophenyl trifluoromethyl sulphide, 2-Bromophenyl trifluoromethyl sulfide
H50576-03
H50576-06
CAAAAH50576-03EA
336.54
CAD
CAAAAH50576-03
CAAAAH50576-06
1-Bromo-2-(trifluoromethylthio)benzene 97%
1-Bromo-2-(trifluoromethylthio)benzene
Purity: 97%
CAS No.: 1644-72-0
Molecular Formula: C7H4BrF3S
Formula Weight: 257.07
Refractive Index: 1.5125
Boiling Point: 92-94°/20mm
MDL No.: MFCD00236347
Formula:
C₇H₄BrF₃S MW: 257.07 g/mol Boiling Pt: 92…94 °C (20 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00236347 CAS Number: 1644-72-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...