4-Chloro-8-methoxy-2-methylquinoline 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50559-03
H50559-06
CAAAAH50559-03EA
210.14
CAD
CAAAAH50559-03
CAAAAH50559-06
4-Chloro-8-methoxy-2-methylquinoline 95%
4-Chloro-8-methoxy-2-methylquinoline
Purity: 95%
CAS No.: 64951-58-2
Molecular Formula: C11H10ClNO
Formula Weight: 207.66
Melting Point: 89-93°
MDL No.: MFCD00272401
Formula:
C₁₁H₁₀ClNO Melting Pt: 89…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00272401 CAS Number: 64951-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...