4-Chloro-7-fluoro-2-methylquinoline 95%

Supplier: Thermo Scientific Chemicals
Warning

H50554-03 H50554-06
CAAAAH50554-03EA 131.3 CAD
CAAAAH50554-03 CAAAAH50554-06
4-Chloro-7-fluoro-2-methylquinoline 95%
4-Chloro-7-fluoro-2-methylquinoline

Purity: 95% CAS No.: 18529-04-9 Molecular Formula: C10H7ClFN Formula Weight: 195.63 MDL No.: MFCD00272333

Formula: C₁₀H₇ClFN
Storage Temperature: Ambient
MDL Number: MFCD00272333
CAS Number: 18529-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR