2-Chloro-3-(trifluoromethyl)quinoxaline 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50549-03
H50549-06
CAAAAH50549-03EA
116.92
CAD
CAAAAH50549-03
CAAAAH50549-06
2-Chloro-3-(trifluoromethyl)quinoxaline 96%
2-Chloro-3-(trifluoromethyl)quinoxaline
Purity: 96%
CAS No.: 254732-51-9
Molecular Formula: C9H4ClF3N2
Formula Weight: 232.59
MDL No.: MFCD00243891
Formula:
C₉H₄ClF₃N₂ Storage Temperature: Ambient |
MDL Number:
MFCD00243891 CAS Number: 254732-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...