4-Chloro-N-methylpicolinamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H50535-03 H50535-MD
CAAAAH50535-03EA 376.04 CAD
CAAAAH50535-03 CAAAAH50535-MD
4-Chloro-N-methylpicolinamide 97%
4-Chloro-N-methylpicolinamide

Purity: 97% CAS No.: 220000-87-3 Molecular Formula: C7H7ClN2O Formula Weight: 170.60 Melting Point: 39-43° MDL No.: MFCD02185921

Formula: C₇H₇ClN₂O
Melting Pt: 39…43 °C
Storage Temperature: Ambient
MDL Number: MFCD02185921
CAS Number: 220000-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR