5-(4-Methoxyphenyl)-2-thiophenecarbaldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

H50483-03 H50483-06
CAAAAH50483-03EA 153.1 CAD
CAAAAH50483-03 CAAAAH50483-06
5-(4-Methoxyphenyl)-2-thiophenecarbaldehyde 99%
5-(4-Methoxyphenyl)-2-thiophenecarbaldehyde

Purity: 99% CAS No.: 38401-67-1 Molecular Formula: C12H10O2S Formula Weight: 218.28 MDL No.: MFCD00663639

Formula: C₁₂H₁₀O₂S
Storage Temperature: Ambient
MDL Number: MFCD00663639
CAS Number: 38401-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR