3,5-Dimethyl-1H-pyrrole-2-carbaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50478-03
H50478-MD
CAAAAH50478-03EA
180.12
CAD
CAAAAH50478-03
CAAAAH50478-MD
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde 97%
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde
Purity: 97%
CAS No.: 2199-58-8
Molecular Formula: C7H9NO
Formula Weight: 123.16
MDL No.: MFCD00111522
Formula:
C7H9NO MW: 123.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00111522 CAS Number: 2199-58-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...