5-(4-Formylphenyl)-1H-tetrazole 99%

Supplier: Thermo Scientific Chemicals
Warning

H50470-03 H50470-MD
CAAAAH50470-03EA 511.92 CAD
CAAAAH50470-03 CAAAAH50470-MD
5-(4-Formylphenyl)-1H-tetrazole 99%
5-(4-Formylphenyl)-1H-tetrazole

Purity: 99% CAS No.: 74815-22-8 Molecular Formula: C8H6N4O Formula Weight: 174.16 MDL No.: MFCD01311033

Formula: C₈H₆N₄O
Storage Temperature: Ambient
MDL Number: MFCD01311033
CAS Number: 74815-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR