4-(4-Fluorophenoxy)benzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H50452-03 H50452-MD
CAAAAH50452-03EA 164.32 CAD
CAAAAH50452-03 CAAAAH50452-MD
4-(4-Fluorophenoxy)benzaldehyde 97%
4-(4-Fluorophenoxy)benzaldehyde

Purity: 97% CAS No.: 137736-06-2 Molecular Formula: C13H9FO2 Formula Weight: 216.21 MDL No.: MFCD01631896

Formula: C₁₃H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD01631896
CAS Number: 137736-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR