3-(3-Methoxyphenyl)-1H-pyrazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50435-03
H50435-06
CAAAAH50435-03EA
158
CAD
CAAAAH50435-03
CAAAAH50435-06
3-(3-Methoxyphenyl)-1H-pyrazole 97%
3-(3-Methoxyphenyl)-1H-pyrazole
Purity: 97%
CAS No.: 144026-74-4
Molecular Formula: C10H10N2O
Formula Weight: 174.20
Melting Point: 90-91°
MDL No.: MFCD01940432
Formula:
C₁₀H₁₀N₂O Melting Pt: 90…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD01940432 CAS Number: 144026-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...