8-hydroxyquinoline-2-carbonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
8-Hydroxyquinoline-2-carbonitrile
H50433-03
H50433-06
CAAAAH50433-03EA
126.87
CAD
CAAAAH50433-03
CAAAAH50433-06
8-hydroxyquinoline-2-carbonitrile 98%
8-hydroxyquinoline-2-carbonitrile
Purity: 98%
CAS No.: 6759-78-0
Molecular Formula: C10H6N2O
Formula Weight: 170.17
Melting Point: 133-136°
UN No.: 3439
MDL No.: MFCD00216717
Formula:
C₁₀H₆N₂O Melting Pt: 133…136 °C Storage Temperature: Ambient |
MDL Number:
MFCD00216717 CAS Number: 6759-78-0 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...