3-(4-Methoxyphenyl)propionyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

H50416-03 H50416-06
CAAAAH50416-03EA 78.68 CAD
CAAAAH50416-03 CAAAAH50416-06
3-(4-Methoxyphenyl)propionyl chloride 99%
3-(4-Methoxyphenyl)propionyl chloride

Purity: 99% CAS No.: 15893-42-2 Molecular Formula: C10H11ClO2 Formula Weight: 198.65 UN No.: 3265 MDL No.: MFCD02258671

Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD02258671
CAS Number: 15893-42-2
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR