1-Naphthylacetyl chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H50386-03
H50386-06
CAAAAH50386-03EA
154.52
CAD
CAAAAH50386-03
CAAAAH50386-06
1-Naphthylacetyl chloride 95%
1-Naphthylacetyl chloride
Purity: 95%
CAS No.: 5121-00-6
Molecular Formula: C12H9ClO
Formula Weight: 204.66
Refractive Index: 1.6210
Odor: Pungent
Boiling Point: 130°/0.6mm
UN No.: 3265
MDL No.: MFCD00175852
Formula:
C₁₂H₉ClO Boiling Pt: 130 °C/0.6 mm Density: 1.23 Storage Temperature: Ambient |
MDL Number:
MFCD00175852 CAS Number: 5121-00-6 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...