(Methylsulfonyl)methanesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

H50385-03 H50385-06
CAAAAH50385-03EA 175.38 CAD
CAAAAH50385-03 CAAAAH50385-06
(Methylsulfonyl)methanesulfonyl chloride 98%
(Methylsulfonyl)methanesulphonyl chloride

Purity: 98% CAS No.: 22317-89-1 Molecular Formula: C2H5ClO4S2 Formula Weight: 192.63 Melting Point: 110° UN No.: 3261 MDL No.: MFCD00052809

Formula: C₂H₅ClO₄S₂
MW: 192.63 g/mol
Melting Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00052809
CAS Number: 22317-89-1
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR