5-Chlorosulfonyl-2-hydroxybenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H50374-03
H50374-06
CAAAAH50374-03EA
146.94
CAD
CAAAAH50374-03
CAAAAH50374-06
5-Chlorosulfonyl-2-hydroxybenzoic acid 97%
5-Chlorosulfonyl-2-hydroxybenzoic acid
Purity: 97%
CAS No.: 17243-13-9
Molecular Formula: C7H5ClO5S
Formula Weight: 236.63
Melting Point: 171-174°
UN No.: 2923
MDL No.: MFCD00052974
Formula:
C₇H₅ClO₅S MW: 236.63 g/mol Melting Pt: 171…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052974 CAS Number: 17243-13-9 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...