4-Amino-N,N-dimethylbenzamide 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H50364-03
H50364-06
CAAAAH50364-03EA
293.88
CAD
CAAAAH50364-03
CAAAAH50364-06
4-Amino-N,N-dimethylbenzamide 97+%
4-Amino-N,N-dimethylbenzamide
Purity: 97+%
CAS No.: 6331-71-1
Molecular Formula: C9H12N2O
Formula Weight: 164.21
Melting Point: 151-153°
MDL No.: MFCD00523648
Formula:
C₉H₁₂N₂O Melting Pt: 151…153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00523648 CAS Number: 6331-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...