4-Amino-7-(trifluoromethyl)quinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

H50355-03 H50355-MD
CAAAAH50355-03EA 233.84 CAD
CAAAAH50355-03 CAAAAH50355-MD
4-Amino-7-(trifluoromethyl)quinoline 98%
4-Amino-7-(trifluoromethyl)quinoline

Purity: 98% CAS No.: 243666-11-7 Molecular Formula: C10H7F3N2 Formula Weight: 212.18 MDL No.: MFCD00269411

Formula: C₁₀H₇F₃N₂
MW: 212.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00269411
CAS Number: 243666-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR