2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethanamine 97%

Supplier: Thermo Scientific Chemicals
Danger

H50333-03 H50333-MD
CAAAAH50333-03EA 418.7 CAD
CAAAAH50333-03 CAAAAH50333-MD
2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethanamine 97%
2-(3,5-Dimethyl-1H-pyrazol-1-yl)ethanamine

Purity: 97% CAS No.: 62821-88-9 Molecular Formula: C7H13N3 Formula Weight: 139.20 UN No.: 2735 MDL No.: MFCD04967722

Formula: C₇H₁₃N₃
Storage Temperature: Ambient
MDL Number: MFCD04967722
CAS Number: 62821-88-9
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR