N-Benzhydryl-N-methylamine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Methylbenzhydrylamine
H50327-03
H50327-06
CAAAAH50327-03EA
77.26
CAD
CAAAAH50327-03
CAAAAH50327-06
N-Benzhydryl-N-methylamine 98%
N-Benzhydryl-N-methylamine
Purity: 98%
CAS No.: 14683-47-7
Molecular Formula: C14H15N
Formula Weight: 197.28
Melting Point: 38-42°
Beilstein Reference: 2096409
MDL No.: MFCD00467853
Formula:
C₁₄H₁₅N MW: 197.28 g/mol Melting Pt: 38…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00467853 CAS Number: 14683-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...