1-Methyl-1H-pyrazole-3-carboxylic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

H50317-03 H50317-MD
CAAAAH50317-03EA 243.32 CAD
CAAAAH50317-03 CAAAAH50317-MD
1-Methyl-1H-pyrazole-3-carboxylic acid 96%
1-Methyl-1H-pyrazole-3-carboxylic acid

Purity: 96% CAS No.: 25016-20-0 Molecular Formula: C5H6N2O2 Formula Weight: 126.11 Odor: Odorless Melting Point: 146-156° MDL No.: MFCD00464254

Formula: C₅H₆N₂O₂
Melting Pt: 146…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00464254
CAS Number: 25016-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR