1-Phenyl-1H-pyrazole-4-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

H50305-03 H50305-MD
CAAAAH50305-03EA 409.22 CAD
CAAAAH50305-03 CAAAAH50305-MD
1-Phenyl-1H-pyrazole-4-carboxylic acid 99%

Purity: 99% CAS No.: 1134-50-5 Molecular Formula: C10H8N2O2 Formula Weight: 188.19 MDL No.: MFCD00514518

Formula: C₁₀H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00514518
CAS Number: 1134-50-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR