N-Boc-1,3-propanediamine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-(3-Aminopropyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(3-Aminopropyl)carbamate, N-Boc-1,3-propanediamine, tert-Butyl (3-aminopropyl)carbamate, N-1-Boc-1,3-diaminopropane·HCl, N-(tert-Butoxycarbonyl)-1,3-propanediamine, N-Boc-1,3-diaminopropane
H50304-03
H50304-06
CAAAAH50304-03EA
152.15
CAD
CAAAAH50304-03
CAAAAH50304-06
N-Boc-1,3-propanediamine 95%
N-Boc-1,3-propanediamine
Purity: 95%
CAS No.: 75178-96-0
Molecular Formula: C8H18N2O2
Formula Weight: 174.24
Melting Point: ca 22°
UN No.: 3259
MDL No.: MFCD00210021
Formula:
C₈H₁₈N₂O₂ MW: 174.244 g/mol Melting Pt: ca 22 °C Storage Temperature: Ambient |
MDL Number:
MFCD00210021 CAS Number: 75178-96-0 UN: 3259 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...